ContactUsLinkComponent
mille2_955x590.jpg
Miele laboratory glassware washers
Miele Professional provides a range of lab washers, reprocessing procedures and coordinated accessories for reprocessing different laboratory glassware different types of laboratory glassware. These solutions help labs achieve reproducible results in complex applications, including organic, inorganic, physical chemistry, biology and microbiology applications in hospital laboratories, pharmacies and the food and cosmetics industries.
Miele’s expertise has led to the development of products available with multiple programs, load carriers, modules and other accessories to meet a variety of specialized requirements. Over the past 60 years, Miele has configured its wash cycles to allow users to select the most appropriate cycle for their glassware. These practical and efficient solutions make Miele a suitable option for laboratory users.
Advantages of Miele
  • Complete documentation of all reprocessing processes to ensure the highest level of safety
  • Maximum flexibility and intuitive use, thanks to new modular baskets and inserts
  • Perfect cleaning performance with ideal water pressure during all program phases due to Miele’s patented variable-speed circulation pump
  • The highest standards of hygiene with Miele’s smoothly welded and cove-cornered chamber and the relocation of heater elements from the chamber to the circulation pump to prevent debris accumulation
  • Individualized solutions, thanks to a variety of customizable features to find perfect solutions for a wide range of applications
Looking for a laboratory glassware washer?
Compact laboratory glassware washers
Laboratory glassware washers from Miele Professional represent commercial-grade solutions for laboratory glassware cleaning in analytical applications.
Miele PG 85 series of undercounter glassware washers
mille_pg85-200.jpg
The new generation of washers/disinfectors for the laboratory.
Watch video
SlimLine laboratory glassware washers
With its new SlimLine laboratory glassware washers, high-performance reprocessing methods, perfectly matching ProCare Lab chemicals and practical accessories, Miele is offering a comprehensive and systematic solution for the reprocessing of laboratory glassware for analytical experiments. Going beyond standard approaches, Miele engineers and users have come up with individualized solutions to tackle everyday laboratory challenges.
Miele SlimLine PLW 6011/6111
Large-capacity laboratory glassware washers
The new PLW 86 model sets new standards in terms of performance and user friendliness. This entirely redesigned washing system boasts considerably better performance compared with that of the previous model series and, thanks to a variable speed pump, can adjust circulation pressure and water consumption to the load in hand.
Miele PLW 86 large-chamber lab washer
Featured products
Have questions about Miele laboratory glassware washers?