Orders LinkContactUsLinkComponent
drive-rapid-flow-1920x760.jpg
MasterSense™ Smart Peristaltic Pumps
Designed for today's critical process applications and the biopharma industry of tomorrow
MasterSense Peristaltic Pumps with Intuitive Touch Screen | Avantor
MasterSense™ monitors your media so you don't have to
With the extra time, a process engineer could:
MasterSense_monitors_your_media_so_you_dont_have_t.png
MF-mastersense-family-connectivity.png
"Monitor your process remotely via PC or mobile device."
(it does not store programs or impact regulatory compliance)
Monitor your process remotely via PC or mobile device
mastersense-peristaltic-pumps-260x190.jpg
Find your Complete Pump System
Masterflex pump Selection Tool quickly steps thru your needs
Start now
More Pumping Options with Advanced Touch-Screen Operation
reglo-multichannel-outlined.png
Cartridges and stopped tubing ensure standard tension in occlusion bed for repeatable performance in low volume/low flow rate pumping.
LOW FLOW
0.0001 to 68 mL/min
*depending on cartridge type
PRESSURE
Up to 35 psi
CONNECTIVITY
EtherNet/IP, analog remote, serial control, plus MasterflexLive
miniflex-digital-dula-channel-pump-with-mf-live-ta.jpg
  • Choose 1 or 2 channels
  • Twist-and-open design, and customizable pump flow patterns ideal for laboratory use.
LOW FLOW
0.004 to 365 mL/min
PRESSURE
Up to 35 psi
CONNECTIVITY
EtherNet/IP, analog remote, serial control, plus MasterflexLive
reglo-gear-outlined.png
Accurate and pulseless flows for pressurized and differential pressure applications and filling.
MEDIUM FLOW
1.7 to 5850 mL/min
PRESSURE
Up to 300 psi (20.7 bar)
CONNECTIVITY
EtherNet/IP, analog remote, serial control, plus MasterflexLive
7801860-ismatec-reglo-digital-piston-pump-drive-mf.jpg
Inert and accurate with touch-screen programmability for high pressure applications and dispensing sensitive or corrosive liquids.
LOW FLOW
0.025 to 75 mL/min
PRESSURE
Up to 100 psi (6.9 bar)
CONNECTIVITY
EtherNet/IP, analog remote, serial control, plus MasterflexLive
touchscreen-icon-100.png
Intuitive and innovative
  • Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
  • Store and recall standard protocols, ensuring continuity
  • Intuitive digital touchscreen with optimized menu navigation and program input - even with gloves on
  • The MasterSense drives offer 3 tiers of access, allowing for varying levels of programming, all password protected, adhering to GMP best practices
certified-icon-100.png
Easy and accurate
  • Operational modes include continuous run, volume dispense, and time dispense
  • Batch control and interval/off time in volume and time modes
  • Qualitative tube calibration ensures tubing is optimized for precise dispensing
  • Graphical indicator lets you know when tubing is optimally calibrated
connectivity-icon-100.png
Connected and smart
  • Over-the-air firmware updates ensure your pump system has the latest software updates, without returning to the manufacturer
  • MasterflexLive® cloud application securely monitors, all MasterSense™ pump systems
  • With MasterflexLive, get notifications to alert you to changes in operating conditions and error messages 24/7
Compare MasterSense Pumps
BT-biopharma-outlined.png
Masterflex® B/T® MasterSense™ BioPharma Process Pump
  • IP66 washdown-rated 316 stainless steel housing
  • Continuous-duty, reversible, variable-flow
  • Choose PerfectPosition® Pump Tubing, B/T 87 or B/T 91
HIGHEST FLOW
0.65 to 42 LPM
PRESSURE
Up to 35 psi
CONNECTIVITY
EtherNet/IP, Profibus, analog remote, serial control, plus MasterflexLive
drive-rapid-flow.png
  • Reliable pumps with the capacity and power for process applications
  • Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000∶1 turndown ratio
  • Add up to two I/P pump heads, or shop Complete Pumps
  • IP66 NEMA 4X stainless steel drives and complete systems
HIGH FLOW
0.001 to 26 LPM
PRESSURE
Up to 40 psi (2.7 bar)
CONNECTIVITY
EtherNet/IP, Profibus, analog remote, serial control, plus MasterflexLive
LS-abs-and-ss-housing.jpg
Masterflex® L/S® MasterSense™ Pumps
  • Small footprint saves on benchtop space
  • Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000:1 turndown ratio
  • Add up to two L/S Pump heads, or shop Complete Pumps
  • IP66 NEMA 4X stainless steel drives and complete systems
MEDIUM FLOW
0.001 to 3400 mL/min
PRESSURE
Up to 150 psi (10.3 bar)
CONNECTIVITY
EtherNet/IP, Profibus, analog remote, serial control, plus MasterflexLive
Pump-Head.png
Masterflex® L/S® MasterSense™ Fill/Finish Pump System With Masterflex® Ultrapharm™ Pump Head
  • All the features of the L/S MasterSense Drive, plus...
  • 250 rpm max speed; dosing volumes from <0.5 mL to >500 mL
  • Whisper quiet operation
  • Offset rollers and planetary gearing ensure dosing accuracy (±0.5%), low pulsation, and long tubing life. Shop Double-Y tubing
MEDIUM FLOW
0.0006 to 19 LPM
PRESSURE
Up to 150 psi (10.3 bar)
CONNECTIVITY
EtherNet/IP or Profibus, plus MasterflexLive or analog remote control
More Pumping Options With Advanced Touch-screen Operation
reglo-multichannel-outlined.png
Cartridges and stopped tubing ensure standard tension in occlusion bed for repeatable performance in low volume/low flow rate pumping.
LOW FLOW
0.0001 to 68 mL/min
*depending on cartridge type
PRESSURE
Up to 35 psi
CONNECTIVITY
EtherNet/IP, analog remote, serial control, plus MasterflexLive
miniflex-digital-dula-channel-pump-with-mf-live-ta.jpg
  • Choose 1 or 2 channels
  • Twist-and-open design, and customizable pump flow patterns ideal for laboratory use.
LOW FLOW
0.004 to 365 mL/min
PRESSURE
Up to 35 psi
CONNECTIVITY
EtherNet/IP, analog remote, serial control, plus MasterflexLive
reglo-gear-outlined.png
Accurate and pulseless flows for pressurized and differential pressure applications and filling.
MEDIUM FLOW
1.7 to 5850 mL/min
PRESSURE
Up to 300 psi (20.7 bar)
CONNECTIVITY
EtherNet/IP, analog remote, serial control, plus MasterflexLive
7801860-ismatec-reglo-digital-piston-pump-drive-mf.jpg
Inert and accurate with touch-screen programmability for high pressure applications and dispensing sensitive or corrosive liquids.
LOW FLOW
0.025 to 75 mL/min
PRESSURE
Up to 100 psi (6.9 bar)
CONNECTIVITY
EtherNet/IP, analog remote, serial control, plus MasterflexLive
mastersense-peristaltic-pumps-260x190.jpg
Find your Complete Pump System
Masterflex pump Selection Tool quickly steps thru your needs
Start now