Orders LinkContactUsLinkComponent
MERCK016721_sq_prove_quickguide_SU_final_04-2020-featured.jpg
Total organic carbon (TOC)
Total organic carbon (TOC) is the amount of carbon bound in an organic compound and is often used as a non-specific indicator of water quality. The rapid colorimetric method of TOC determination is easy and accurate.
  • Rapid digestion
  • Choice of photometers and spectrophotometers and ready-to-use TOC tubes and powder pillows.
Total organic carbon (TOC) | Avantor
toc_solutions.jpg
Detecting organic contamination in water can be simple with total organic carbon (TOC) analysis solutions from Avantor®. Using sodium persulfate as the primary reagent, dissolved organic carbon is oxidized, and the resulting CO₂ is measured with precision using spectrophotometry. Depending on the water type, additional reagents such as acids or bases may be required for accurate analysis. 
VWR® offers a full range of high-quality reagents to support your TOC analysis, including:
  • Sodium hydroxide 1.2 N solution  
  • Sulfuric acid 1.8 N solution  
  • Sodium persulfate in various concentrations  
  • Phosphoric acid (10% and 25% solutions)  
See our full selection of solutions tailored to your specific water testing needs.
Contact us today for more information
toc_merck_tubes.jpg
Merck TOC tubes
You can conduct hassle-free analysis with our user-friendly system. Nearly all the reagents you’ll need come pre-packaged in convenient cells, ready for you to use. If you need additional reagents for determination or sample pre-treatment, we can provide them in easy-to-handle, dose-metering bottles for precise application.
  • Validated and standard-compliant analysis reagents 
  • Bar code system enables rapid and simple operation
  • Comprehensive quality control documentation provided for every test
  • Batch and quality certificates available online
Product selection
MERCK016721-sq-prove-quickguide-SU-final-04-2020-200.jpg
Get streamlined and accurate photometric determination with ready-to-use reagents and cells with Spectroquant® TOC cell tests. Spectroquant® Prove instruments offer a photometric method for quickly and accurately detecting TOC. These instruments automatically recognize Spectroquant® test kits to ensure that the correct analysis method is used and accurate results are displayed.
dp_spectroquant_tr.jpg
You can generate fast, reliable results in chemical oxygen demand (COD) and total organic carbon (TOC) testing with Spectroquant® thermoreactors from Merck. The Spectroquant® TR series provides reliability, speed, precision and future compatibility combined with the simplest possible operation and highest user safety.
Avantor® offers three models of Spectroquant® TR series thermoreactors:
  • TR 320: basic model for 12 cuvettes, with 7 pre-stored temperature programs
  • TR 420: accepts 24 cuvettes and offers an additional 8 user-defined programs 
  • TR 620: accepts 2×12 cuvettes in 2 temperature zones, with 7 pre-stored and 8 user-defined programs
Need more information about our total organic carbon products?