Orders LinkContactUsLinkComponent
battery-science-metal-trace-analysis-1920x760.jpg
Metal trace analysis (High purity acids, AAS and ICP Standards)
In metal analysis, elemental evidence is now gathered from the tiniest traces in the ppm, ppb and ppt ranges with sophisticated instrumentation like AAS, ICP-OES or, more recently, ICP-MS.
Metal trace analysis | Avantor
Product selection
High-purity-teaser.jpg
A complete range of High purity acids with max. permitted levels contaminants in ppb and ppt, essential in ultra-trace analysis applications.
ICP-Standards-teaser.jpg
Complete range of Multi-element solutions for using as internal standard or for a perfect equipment calibration, traceable to NIST and delivered with CoA.
ICPS-TEASER.jpg
Wide range of ICP and ICP-MS Standards engineered to be stable, compatible, traceable to NIST SRMs, and are manufactured and tested under ISO 17034 & ISO 17025 guidelines. Certified values are based on two independent methods, uncertainties are based...