Orders LinkContactUsLinkComponent
Facility-Management.jpg
Advanced Battery Science and Technology Facility Management
Avantor® sets advanced battery science in motion to create a better world. We offer a broad range of products to keep the facility clean, safe and ordered.
Advanced Battery Science and Technology Facility Management | Avantor
Humidity Control
Humidity-Control.jpg
Data loggers and Monitoring sytems of CO2, temperature, humidity and particulate.
Data loggers automatically record precise measurements even when personnel cannot be present.
Monitor temperatures inside rooms, environmental chambers, or cleanrooms.
Systems designed to monitor exposure and cross-contamination.
Mats and Flooring
Mats-and-Flooring.jpg
Floor and Tacky mats for personnel comfort and contamination-free environments. Available in different versions to suit every need.
Mats designed to stimulate the body to increase blood circulation and alertness.
Multi-layered contamination control mats remove and contain direct and dust.
Mats designed to dissipate static from conductive items placed on it.
Mats suction to the ground to prevent shifting while trapping air in its chambers.
Mats with tacky surfaces that remove impurities from shoe soles before stepping into cleanrooms.
Waste Disposal and Recycling
Waste-Disposal.jpg
Disposing and Recycling with an eye on sustainability. Look for the green leaf to explore the range of Environmentally preferable products.
Solution which includes the storage, shipping and recycling of a particular type of waste that isn’t currently recycled through local councils or traditional recycling facilities.
Wide selection of waste containers to choose based on your space and needs.
Conductive ESD waste baskets remove the static threat of plastic receptacles that can damage sensitive electronic devices.
Offered in hygienic and non-sterile varieties, the supplies are created from tough materials to last, and heavy walls give superior tear protection.
Safety Signs and Floor Marking
smoking.jpg
A wide range of signs and tapes to communicate with employees and visitors. Assure protection of equipment and personnel with the choice of padlocks and electrical and valve lockout kits.
Brightly colored biohazard, radiation and high voltage signs warn of the presence of dangerous materials.
Fire extinguisher, alarm and exit signs in addition to first aid kit and AED signs that are essential in an emergency.
Folding stand and cone-shaped caution signs are easy to move and transport, while wall mount "V" shaped signs offer permanent visibility.
A large selection of floor signs alert and remind those in and around walkways, work areas and entrances with bold graphics and messages.
Mark specific areas in the workplace to keep clear of obstructions and debris, designate storage locations and more.
Provide a safe step area by installing slip-resistant tape to stairways, walkways and aisleways.
Keeps equipment inoperative while repairs are being made. Control cannot be turned on until last workers padlock is removed.
Keyed padlocks of aluminum and steel offer lightweight and compact security in a variety of colors.
Lockout boxes are available in red or yellow. They are UL validated for recycled content, made in USA and high quality.
Nonconductive lockout hasps offer exceptional electrical insulating characteristics and are made of orange nylon material.
The electrical lockout toolbox provides all the basics to lock out breakers, electrical plugs, wall switches and fuses.
Striped or solid pressure sensitive tape for delineating and color coding specific equipment and areas.
Lockout Tag Out
Lockout.jpg
Assure protection of equipment and personnel with the choice of padlocks, lockout hasps, electrical and valve lockout kits.
Keeps equipment inoperative while repairs are being made. Control cannot be turned on until last workers padlock is removed.
Keyed padlocks of aluminum and steel offer lightweight and compact security in a variety of colors.
Spill Control
Spill-Control.jpg
Choose from a variety of spill kits to keep your laboratory or facility space clean of spilled chemical solutions.
VWR Spill Kit, Chemical
Hazmat spill response kit includes 20 pads, three socks, one pair nitrile gloves, one pair safety goggles and one water-resistant yellow bag with tie.
Includes PPE to guard against corrosive battery acid. 20-gallon overpack salvage drum container meets DOT shipping specifications for overpacking hazardous materials.
Kit contents are formulated specifically for absorbing and neutralizing battery acid.
Effective designs absorb liquids quickly to leave work areas clean and dry for improved safety.
The granules are suitable for absorbing acids, alkalis, other aqueous solutions and organic liquids.
Eco Spill Kit
Premium absorbent made of 100% recycled material. Extremely effective on the absorption of hydrocarbons, water-based fluids and most aggressive chemicals.
Hand Hygiene and Janitorial Supplies
Hand-Hygiene.jpg
Everything for the hygiene of personnel, equipment and surfaces
Excellent for cleaning rough, abrasive, or irregular surfaces, yet soft enough for delicate surfaces.
These all-purpose wipes are versatile cloth-like wipes that are safe for skin contact with no rinse required.
White PVC dispensers for tissue boxes. Can be wall-mounted or placed on a bench.
Vileda Professional-FHP PVAmicro Cloth
The PVA impregnation makes the cloth very easy to rinse and wring. PVAmicro achieves a perfect, shiny cleaning result without streaks every time.
For reliable performance in general-purpose uses.
Concentrated, all-purpose cleanroom detergent is designed for use between disinfectant applications and for residue removal.
Mild formulation that is designed for frequent hand washing.
Touch-free soap dispensing unit effectively reduces cross-contamination and waste in the cleanroom and gowning areas.
VWR® MicroDispenser™ Touch-Free Soap Dispenser
Touch-free soap dispensing unit effectively reduces cross-contamination and waste in the cleanroom and gowning areas.
First Aid
First-Aid.jpg
Eye Station and First Aid kits for a prompt treatment of injuries and emergencies
Eye flushing solution for irrigating the eyes and eye strips to keep eye injuries protected.
Printers and Labels
Printers-and-Labels.jpg
A wide range of printers to create your labels from lab to the facility, or where you need with the portable models.
Get fast and efficient precision printing for high-mix / high-volume labeling. Printer and software allow for quick label set up and no-waste material.
High-quality labels to enhance safety and general organization in the laboratory or other areas in your facility.
Chemical Handling and Storage
Chemical-Handling.jpg
See our large selection chemical handling and storage products for your laboratory.
Storage cabinets are specifically designed for the storage of chemicals, flammable and combustible liquids.
Closed-system design ensures the safety of your environment and employees at all phases of chemical handling.
ESD Control
ESD-Control.jpg
Apparel and accessories for ESD grounding.
Wearable grounders are made of pliable, adjustable materials that maximize contact to provide consistent grounding.
ESD apparel manufactured from breathable and comfortable polyester and carbon fabric.