Orders LinkContactUsLinkComponent
Spectroscopy_Hero.jpg
Spectroscopy
Find a wide and growing range of spectrophotometers and biophotometers adapted to your needs. Also procure others instruments, reagents, cuvettes etc. for all your associated applications.
Explore more
Spectroscopy | Avantor
Spectroscopy_CardOne.png
Calibration kit consisting of three gray glass filters to verify absorbance accuracy and a holmium oxide glass filter to verify wavelength accuracy.
Spectroscopy_CardTwo.png
VWR offer a total portfolio of VIS, UV/VIS, fluorescence and small volume spectrophotometers. Optical systems include single beam, split beam or double beam. Light source is either tungstem or xenon lamps. Spectrophotometers for life science or water analysis applications are also available.
Spectroscopy_CardThree.png
The Genesys spectrophotometers provide exceptional performance. Choose between VIS and UV/VIS spectrophotometers with single beam, split beam or double optical systems. Light sources can be either tungsten or xenon lamps.
Find a wide and growing range of spectrophotometers and biophotometers adapted to your needs. Also procure others instruments, reagents, cuvettes etc. for all your associated applications.