ContactUsLinkComponent
FLEXEL CUB.,TPE, MAG.MIX, 200L, 2pcs Tubing: Clear C-Flex 374, Silicone 1 * 2 ST
Catalog # SARTFMB114893
Supplier:  Sartorius
New Product
FLEXEL CUB.,TPE, MAG.MIX, 200L, 2pcs Tubing: Clear C-Flex 374, Silicone 1 * 2 ST
Catalog # SARTFMB114893
Supplier:  Sartorius
New Product
Frequently Bought Together