CTD-Diver 200m, 0-120mS/cm Sensor and logger unit for measuring and recording groundwater level/temperature/conductivity. Memory for 144000 measurements (and 144000 backup). Conductivity 0-120 mS/cm, compensation temperature 0 till +50 C. Measuring 1 * 1 ST
Artikelnr. EIJK11116004
Lieferant: EIJKELKAMP AGRISECTOR
Frequently Bought Together