ContactUsLinkComponent
Embedding Mold, 5 round, numbered cavities, 17.5mm x 2.0mm, each 1 * 1 ST
Artikelnr. SCIAE70929
Lieferant:  SCI MUENCHEN
Embedding Mold, 5 round, numbered cavities, 17.5mm x 2.0mm, each 1 * 1 ST
Artikelnr. SCIAE70929
Lieferant:  SCI MUENCHEN
Frequently Bought Together