Bundhose Multisecuro ASF-1 56 OBERSTOFF:54/45/1% MODACRYL/ BAUMWOLLE/ANTISTATIC,320g/m² Farbe:MARINE/ROYALBLAU Waschbar:bis 60°C,Industriewäschefähig EN-NORMEN: EN11611,Klasse 1,EN11612,A1, B1,C1,E2,F1,EN61482-1-2,Klasse1,EN1149-5 1 * 1 ST
Catalog # RENT4444/56
Supplier: RENTEX
New Product
Frequently Bought Together