Mikroskop-Deckgläser, 24x70 mm, Borosilikatglas D 263® M hickness No. 1 (0,13-0,16 mm), CE-gekennzeichnet, 100 Stück /Scharnierdeckel-Kunststoffbox, 1000 Stück/Karton, 25.000 Stück/Multipack, MARIENFELD SUPERIOR Microscope cover glasses, 24x70 mm, bo 1 * 1.000 ST
Artikelnr. MARI0101992091
Lieferant: MARIENFELD
New Product
Frequently Bought Together