ContactUsLinkComponent
48-well FlowerPlate MF MTP with flower-shaped wells and transparent bottom with integrated microfluidics for feeding and pH control of 32 cultivation wells (16 reservoir wells), Gen 2, pH and DO optodes type 2 (LG1/RF), 1 box (1 MTP). 1 * 1 SET
Artikelnr. BECKM2PMTPMF32CBOH
Lieferant:  Beckman Coulter
New Product
48-well FlowerPlate MF MTP with flower-shaped wells and transparent bottom with integrated microfluidics for feeding and pH control of 32 cultivation wells (16 reservoir wells), Gen 2, pH and DO optodes type 2 (LG1/RF), 1 box (1 MTP). 1 * 1 SET
Artikelnr. BECKM2PMTPMF32CBOH
Lieferant:  Beckman Coulter