AuftragsverfolgungKontakt
Antibiotic Sensitivity Discs | Avantor
Antibiotic Sensitivity Discs
Ceftizoxime, 30 mcg, 50
Artikelnr. HIMESD110-5X50DS
Anbieter:   HIMEDIA
Antibiotic Sensitivity Discs
Artikelnr. HIMESD110-5X50DS
Anbieter:  HIMEDIA
CAS Nummer:  
Antibiotic Sensitivity Discs
Ceftizoxime, 30 mcg, 50
CAS Nummer:  
Preis
19,50 €
Einloggen für Verfügbarkeit
1 * 50 ST
Product Details & Documents
Antibiotic sensitivity disc are used to determine the effectivness of an antibiotic against a microorganism as a guide to antibiotic therapy.
The disc consist of a specific concentration of an antibiotic in mcg which is tested against a microorganism of a standard density, swabbed uniformly on an appropriate media following overnight incubation.
The antibiotic is impregnated on a filter paper disc in specified concentration. Both sides of the disc are printed with name and concentration for easy identification.
Spezifikationen
Spezifikationen
  • VE:
    50
  • Bezeichnung:
    Ceftizoxime, 30 mcg
Spezifikationen