ContactUsLinkComponent
Microscope slides, 13x11+0/-0,4 mm, thickness 1 mm, white glass, cut edges, without frosted end, in boxes of 50 pcs., 500 pcs./carton, paper-interleaved, CE marked, MARIENFELD SUPERIOR 1 * 1 ST
Artikelnr. MARI11300900928
Lieferant:  MARIENFELD
New Product
Microscope slides, 13x11+0/-0,4 mm, thickness 1 mm, white glass, cut edges, without frosted end, in boxes of 50 pcs., 500 pcs./carton, paper-interleaved, CE marked, MARIENFELD SUPERIOR 1 * 1 ST
Artikelnr. MARI11300900928
Lieferant:  MARIENFELD
New Product
Frequently Bought Together