ContactUsLinkComponent
Minivette® POCT EDTA K3E, nominal volume: 200 µl, preparation: K3 EDTA, violet, colour code ISO, 150 piece(s)/bag, 150 piece(s)/case 1 * 1.500 ST
Catalog # SARS17.2113.201
Supplier:  SARSTEDT
Minivette® POCT EDTA K3E, nominal volume: 200 µl, preparation: K3 EDTA, violet, colour code ISO, 150 piece(s)/bag, 150 piece(s)/case 1 * 1.500 ST
Catalog # SARS17.2113.201
Supplier:  SARSTEDT
Frequently Bought Together