ContactUsLinkComponent
HD-Lite camera, color HD High definition 5 MP high speed camera 1080p with HD screen. 24 bit color rendering, standard SD card, HDMI and USB-2 output with built-in capture software. Can be used in stand alone or with a computer and supplied with Imag 1 * 1 ST
Artikelnr. EUMXVC.3032-HDS
Lieferant:  EUROMEX
New Product
HD-Lite camera, color HD High definition 5 MP high speed camera 1080p with HD screen. 24 bit color rendering, standard SD card, HDMI and USB-2 output with built-in capture software. Can be used in stand alone or with a computer and supplied with Imag 1 * 1 ST
Artikelnr. EUMXVC.3032-HDS
Lieferant:  EUROMEX
New Product
Frequently Bought Together