Express Eingabe
Austria
AuftragsverfolgungKontakt
ReadiPrep™ Exosome Isolation Kit | Avantor
ReadiPrep™ Exosome Isolation Kit
200 preparations, 200
Artikelnr. AATB60205
Anbieter:   AAT Bioquest
ReadiPrep™ Exosome Isolation Kit
Artikelnr. AATB60205
Anbieter:  AAT Bioquest
CAS Nummer:  
ReadiPrep™ Exosome Isolation Kit
200 preparations, 200
CAS Nummer:  
Preis
620,00 €
Einloggen für Verfügbarkeit
1 * 200 ST
Product Details & Documents
Designed for consistent and dependable results. The ReadiPrep™ exosome isolation kit offers a streamlined and effective approach to isolating intact exosomes from diverse biological samples, including cell culture media and serum.
  • Robust protocol
  • Produced exosomes are suitable for further downstream analyses or purification via affinity-based methods
  • Six-month shelf life
The principle behind this kit is based on the selective precipitation of exosomes by inducing the removal of water molecules from the solution.
Lieferumfang: Shipped at RT.
Achtung! Bitte beachten: Research use only.
Spezifikationen
Spezifikationen
  • VE:
    200
  • Pk Info:
    200 preparations
Spezifikationen