Lysates, HeLa (human)
Lysate, HeLa, human, heat shocked, with no recovery time, Pooled, 200 µG
Catalog # ENZOADILYCHL102F
Supplier: Enzo Life Sciences
Jump to:
Product Details & Documents
The HeLa cell line derived from a human donor's epithelioid carcinoma tissue (cervix).
Dilution range: 20 µg/lane (Western blotting)
Liquid lysates in SDS-PAGE sample buffer.
Heat shock-treated HeLa cells with no recovery time can be a useful tool in studying the regulators of immediate-early gene expression during heat shock stress response.
Specifications
Specifications
- VE:200 µG
- Einzelspender/Pool:Pooled
- Bezeichnung:Lysate, HeLa, human, heat shocked, with no recovery time
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.