AuftragsverfolgungKontakt
Microarray-Objektträger, MULTIVIEW®
Microarray-Objektträger, MULTIVIEW®
Universal tissue microarray slides, 50 slides
Catalog # ENZOADI9500200050
Supplier:   Enzo Life Sciences
Microarray-Objektträger, MULTIVIEW®
Catalog # ENZOADI9500200050
Supplier:  Enzo Life Sciences
Microarray-Objektträger, MULTIVIEW®
Universal tissue microarray slides, 50 slides
Price
1 560,00 €
Log in for availability
1 * 5 ST
Product Details & Documents
These microarray slides with eleven human tissue specimens includes placenta, liver, tonsil, colon, skin, brain, breast (fibroadenoma), prostate, thyroid, kidney and fallopian tube.
  • High-consistency specimen and slide preparation for reduced maintenance
  • Fixed in formalin and paraffin embedded
  • Sections are placed on a SuperFrost Plus slide
  • Cores are approximately 2 mm in diameter
  • Tissue sections are 3 µm thick
Suitable for use with common FFPE tissue protocols as rapid screening tools for the optimisation or validation of stains in a broad-spectrum of immunohistochemical procedures.
Specifications
  • Bezeichnung:
    Universal tissue microarray slides, 50 slides
  • Seq:
    06
  • Cat. No.:
    ENZOADI9500200050