AuftragsverfolgungKontakt
Extraction reagents
Extraction reagents
Extraction reagent 1, 5X, 10 mL
Catalog # ENZOADI801526
Supplier:   Enzo Life Sciences
Extraction reagents
Catalog # ENZOADI801526
Supplier:  Enzo Life Sciences
CAS Number:  
Extraction reagents
Extraction reagent 1, 5X, 10 mL
CAS Number:  
Price
175,00 €
Log in for availability
1 * 10 mL
Product Details & Documents
Concentrated lysis buffer for preparation of cell and tissue extracts.
TRIS buffer containing detergents.
Specifications
Specifications
  • VE:
    10 mL
  • Bezeichnung:
    Extraction reagent 1, 5X