Express Eingabe
Austria
AuftragsverfolgungKontakt
VHB DNA Washing Buffer
VHB DNA Washing Buffer
VHB buffer, 1.760 mL
Catalog # OMEGVHB-1760
Supplier:   OMEGA BIO-TEK
VHB DNA Washing Buffer
Catalog # OMEGVHB-1760
Supplier:  OMEGA BIO-TEK
CAS Number:  
VHB DNA Washing Buffer
VHB buffer, 1.760 mL
CAS Number:  
Price
2 310,00 €
Log in for availability
1 * 1.760 mL
Product Details & Documents
VHB buffer must be diluted with ethanol before use.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
  • VE:
    1.760 mL
  • Bezeichnung:
    VHB buffer
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.