Pass through cabinets
Pass through cabinet, with static dissipative window, sealing gasket and compression handle, 2, 700×600×700 mm
Artikelnr. PALBPT-706070MS
Anbieter: Palbam
Springe zu:
Product Details & Documents
Pass Through cabinet allows products and materials to be transferred between different rooms. Reduces unnecessary walking and entering/exiting cleanroom.
- Single or double versions available
- Sealing gaskets and compression handles ensure firm closure
- Tempered Glass or anti static PVC windows available
- Mechanical or electronic interlocks available
- Can be customised to requirements
Fabrication material: 304 stainless steel with electropolished finish.
Dokumente
Palbam Class - Catalog 2018
Produktliteratur
Spezifikationen
- Bezeichnung:Pass through cabinet, with static dissipative window, sealing gasket and compression handle
- Seq:02
- B×T×H:700×600×700 mm
- Türen:2