ContactUsLinkComponent
C25Z-2143EUHB - 3 mpos dead-end path, u-electric, USB 1/16*.4mm, 50C/250psi liq, PEEK E4/E Universal electric actuator Serial interface: USB 1 * 1 ST
Artikelnr. VICIC25Z-2143EUHB
Lieferant:  VICI
C25Z-2143EUHB - 3 mpos dead-end path, u-electric, USB 1/16*.4mm, 50C/250psi liq, PEEK E4/E Universal electric actuator Serial interface: USB 1 * 1 ST
Artikelnr. VICIC25Z-2143EUHB
Lieferant:  VICI
Frequently Bought Together