AuftragsverfolgungKontakt
Codesoft Lite Software | Avantor
Codesoft Lite Software
Codesoft lite subscription online
Artikelnr. BRDY196181
Anbieter:   Brady
Codesoft Lite Software
Artikelnr. BRDY196181
Anbieter:  Brady
Codesoft Lite Software
Codesoft lite subscription online
Preis
441,00 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Codesoft Lite label design software can be used for asset tracking, barcode labelling, and inventory and inspection labelling applications.
  • Only available as a single user license
  • Limited to 1 thermal printer
Languages availble include Arabic (Egypt), Czech, Danish, Dutch, English, Finnish, French, German, Hebrew, Hungarian, Italian, Norwegian (Bokmål), Polish, Portuguese, Romanian, Russian, Spanish, Swedish, and Turkish.
Compatible with BBP11, BBP12, BBP72, PR Plus, IP Series, PAM3000, PAM3600, THT 76mm Core, and TLS-PC LINK printers.
Spezifikationen
  • Bezeichnung:
    Codesoft lite subscription online
  • Seq:
    01