Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Cerex® TRACE-N SPE Columns

Column Cerex TRACE-N, 10 µm, 1 ml, 20 mg
  TCAT610-0201R
 :   Tecan
Cerex® TRACE-N SPE Columns
  TCAT610-0201R
 :  Tecan
Cerex® TRACE-N SPE Columns

Column Cerex TRACE-N, 10 µm, 1 ml, 20 mg

 
 
   
1 * 96 ST
 

End-capped C18 chemistry.

  • Reversed-phase
  • Liquid handler compatible

A 10 µm micro particulate with end-capped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep.

 
  • Partikelgröße:
    10 µm
  • Kapazität:
    1 ml
  • Bezeichnung:
    Column Cerex TRACE-N
  • Seq:
    03
  • Bettgewicht:
    20 mg
  • Cat. No.:
    TCAT610-0201R
  • Endcapped:
    Ja
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.