[EN]10 COMPONENTS IN CHLOROFORM 1 * 5 Ampull
Catalog # CMPLRD0970702.5
Supplier: CUSTOM MADE CHEMICALS LAB
[EN]10 COMPONENTS IN CHLOROFORM 1 * 5 Ampull
Catalog # CMPLRD0970702.5
Supplier: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Schnelltests, Gesamthärte-Test, MQuant®, Supelco®
Supplier: Merck
Testsätze, Ammonium, Spectroquant®, Supelco®
Supplier: Merck
Ammoniak-Lösung 25%, LiChropur™ für die HPLC, Supelco®
Supplier: Merck
Testsätze, Chlor-Test, Spectroquant®, Supelco®
Supplier: Merck
Testsätze, Eisen-Test, Spectroquant®, Supelco®
Supplier: Merck
Natriumhydroxid ≥97,0%, Perlen, TECHNICAL
Supplier: VWR Chemicals
Acid Red 183 30% (Farbstoffgehalt), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
TRITON X100/ SULPHURIC ACID MIX 1 * 10 L
Supplier: CUSTOM MADE CHEMICALS
BIOTECTOR 1.8N SCHWEFELSAURE 80MG/L MN 1 * 20 L
Supplier: Hach
Acid red 52, technische Qualität, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Acid red 27 ≥85-95% (Farbstoffgehalt), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Rhodamin B ≥95% (durch HPLC), Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Testkits, photometrisch, Ammonium, LCK, Küvettentest
Supplier: Hach
Fast Red RC Salt, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Powder pillows, ammonia
Supplier: Hach