DP23M-CU-1-2 monochrome camera 6.4 Mpx, 1/1.8 inch, backside illuminated monochrome CMOS Camera for C-mount adaption, no cooling, USB 3.1 interface. Including: camera head, USB 3.1 cable and safety instructions. Supported by cellSens and DP2-TWAIN 1 * 1 ST
OLYME9701759
Frequently Bought Together