SULFUR 20µG/G IN ISOOCTANE 1 * 100 mL
Artikelnr. ACCUSTP-2X-100ML
Lieferant: AccuStandard
Frequently Bought Together
Osmolalitäts standards
Lieferant: VWR Chemicals
FENOXAPROP-P (R-ENANTIOMER) CERTIFIED 1 * 0,1 g
Lieferant: EHRENSTORFER
CYCLIZINE-13C6 HCL 1 * 1 mL
Lieferant: Merck
TERBINAFINE RELATED COMPOUND B 1 * 50 mg
Lieferant: Merck
AMINOPYRALID CERTIFIED 1 * 100 mg
Lieferant: EHRENSTORFER
Riboflavin (Vitamin B2)
Lieferant: EDQM
[EN]SODIUM CHLORIDE 99.995% ISO 17035 17 1 * 50 g
Lieferant: CPACHEM
Magnesium standard solution, for ICP, in 5% HNO₃, VHG
Lieferant: LGC Standards PROMOCHEM
Pharmaceutical secondary standard, Fructose, (CRM), Supelco®
Lieferant: Merck
NEPODIN 1 * 5 mg
Lieferant: Merck
Quinine Sulfate, Reference Standard
Lieferant: USP
Supelco® PESTANAL® Organic Reference Standard, Methomyl
Lieferant: Merck
pH buffer, single-use, SINGLET™
Lieferant: Hach
FONOFOS CERTIFIED 1 * 0,1 g
Lieferant: EHRENSTORFER
Triethyl Citrate, Reference Standard
Lieferant: USP