ContactUsLinkComponent
with 10 removable rolls of PVC of 355 mm length, 10-70 r.p.m., power supply with connectors for EU,US, UK and AU, 24 V DC 1 * 1 ST
Catalog # MARI7202037
Supplier:  MARIENFELD
with 10 removable rolls of PVC of 355 mm length, 10-70 r.p.m., power supply with connectors for EU,US, UK and AU, 24 V DC 1 * 1 ST
Catalog # MARI7202037
Supplier:  MARIENFELD
Frequently Bought Together