AuftragsverfolgungKontakt
Bioanalyzer High Sensitivity RNA Analysis | Avantor
Bioanalyzer High Sensitivity RNA Analysis
Agilent RNA 6000 Pico reagents, Reagents only, no chips included, sufficient for 25 chips
Artikelnr. AGLS5067-1514
Anbieter:   AGILENT
Bioanalyzer High Sensitivity RNA Analysis
Artikelnr. AGLS5067-1514
Anbieter:  AGILENT
Bioanalyzer High Sensitivity RNA Analysis
Agilent RNA 6000 Pico reagents, Reagents only, no chips included, sufficient for 25 chips
Preis
538,94 €
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
High sensitivity RNA sample quality assessment.
  • Maximum flexibility is enabled with options to check total RNA, mRNA, or cRNA quality
  • Minimal sample consumption required with only need 50 pg of total RNA for an effective analysis
  • Easily determine the RNA integrity number (RIN) using in-built software that automatically assigns an integrity number to a total RNA sample
  • This tool was designed to provide scientists an objective measurement of degradation in RNA samples
  • RNase free consumables avoid sample degradation during analysis
  • Accurate results are achieved rapidly, with 11 samples analysed in approximately 30 minutes
The Bioanalyzer RNA 6000 Pico assay provides reliable and reproducible characterisation of total RNA and mRNA from multiple sample types, from as little as 50 pg of total RNA. The software provides an objective measurement of RNA quality with RIN (RNA Integrity Number). Validated RIN values are available for eukaryotic, prokaryotic, and plant samples.
The Bioanalyzer RNA kits allow dependable and precise integrity checks and sample quantitation prior to any RNA-dependent application. This ensures the data from downstream research processes, such as gene expression microarray, qPCR, and transcriptome sequencing, are valid and reliable.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Spezifikationen
  • Bezeichnung:
    Agilent RNA 6000 Pico reagents
  • Seq:
    02
  • Details:
    Reagents only, no chips included, sufficient for 25 chips