[EN]MERCURY (I) NITRATE 0.1M (0.1N) 1 * 500 mL
Artikelnr. CMPLZ14031040
Lieferant: CUSTOM MADE CHEMICALS LAB
[EN]MERCURY (I) NITRATE 0.1M (0.1N) 1 * 500 mL
Artikelnr. CMPLZ14031040
Lieferant: CUSTOM MADE CHEMICALS LAB
Frequently Bought Together
Sol.,Per Acid 0.5%,500ML 1 * 1 ST
Lieferant: Epredia
[EN]6 COMPONENTS IN HNO3 2% 1 * 100 mL
Lieferant: CUSTOM MADE CHEMICALS LAB
Solvent black 5, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Reagents for Phosphate Determination using Lovibond® MD photometers
Lieferant: Lovibond
Zincon sodium salt ≥75% (Farbstoffgehalt), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
5-Fluoruracil ≥99% (durch HPLC), Pulver, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
trans-4-Hydroxyzimtsäure ≥98% (durch HPLC), Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
[EN]7 COMPONENTS EACH IN HNO3 10% 1 * 100 mL
Lieferant: CUSTOM MADE CHEMICALS LAB
[EN]7 COMPONENTS 1000MG/L EACH 1 * 5 Ampull
Lieferant: CUSTOM MADE CHEMICALS LAB
50 TABLETTEN MOLYBDATE LR N2 PHOTOMETER 1 * 50 ST
Lieferant: WATER ID PRIMELAB