L(-)-Prolin ≥99%
Bottle, 100 g
Artikelnr. APOSBIP0717-100G
Anbieter: Apollo Scientific
Springe zu:
Product Details & Documents
USP, pyrogen free
Synonyme: (S)-(-)-ProlineH-Pro-OHL-Proline, H-Pro-OH
- CAS Nummer:147-85-3
- Formel:C₅H₉NO₂
- MDL-Nummer:MFCD00064318
- Molekulargewicht:115.13 g/mol
- Schmelzpunkt:228 °C (Zersetzung)
- Dichte:1,36 g/cm³ (25 °C)
- Lagertemperatur:Raumtemperatur
- EINECS:205-702-2
Documents
Zertifikate
Suche nach ChargennummerSpezifikationen
Spezifikationen
- Packung:Bottle
- VE:100 g
Spezifikationen
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



