REINRAUM MOPP KING CR TUCHMOPP 40 CM - gamma irradiated - disposable mop, autoclavable - for pre-preparation - up to ISO class 5 - for sterile areas (GMP class A/B) - pre-washed - single packed - double wrapped - 10 mop packed in LDPE bag - gamma irr 1 * 10 ST
Catalog # PPSA3500170
Lieferant: PFENNIG REINIGUNGSTECHNIK
Frequently Bought Together