Express Eingabe
Austria
AuftragsverfolgungKontakt
WHEATON® COMPLETEPACK Sterile Components, Stoppers Only | Avantor
WHEATON® COMPLETEPACK Sterile Components, Stoppers Only
Omni flex sterile igloo lyo stopper, 20 mm Ø
Artikelnr. WHEAW988008-S
Anbieter:   DWK Life Sciences
WHEATON® COMPLETEPACK Sterile Components, Stoppers Only
Artikelnr. WHEAW988008-S
Anbieter:  DWK Life Sciences
WHEATON® COMPLETEPACK Sterile Components, Stoppers Only
WHEATON® COMPLETEPACK Sterile Components, Stoppers Only
Omni flex sterile igloo lyo stopper, 20 mm Ø
Preis
830,00 €
Einloggen für Verfügbarkeit
1 * 125 ST
Product Details & Documents
  • Sterilised, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Complete manufacturing and certificate of analysis traceability
  • Ready-to-use, Just-in-time, minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
 : USP 788 / EP 2.9.19 Particulate Matter in Injections (Vials and Stoppers), USP 85/ EP 2.6.1, The Bacterial Endotoxins Test (Vials and Stoppers) and lot certified to SAL 10⁻⁶ (Vials, Stoppers, and Seals), USP 660 / EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381 / EP 3.2.9 Elastomeric Closures used in Parenteral Preparations.
Spezifikationen
  • Bezeichnung:
    Omni flex sterile igloo lyo stopper, 20 mm Ø
  • Seq:
    06