Express Eingabe
Austria
AuftragsverfolgungKontakt
 

Tyloxapol USP

Glass, amber, 5 g
Catalog # SPCMT1708-5GM
Supplier:   Spectrum Chemical
Tyloxapol USP
Catalog # SPCMT1708-5GM
Supplier:  Spectrum Chemical
CAS Number:  25301-02-4
Tyloxapol USP

Glass, amber, 5 g

CAS Number:  25301-02-4
Price
424,00 €
Log in for availability
1 * 5 g
Product Details & Documents

All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

  • CAS Number:25301-02-4
  • MDL Number:MFCD00149002
  • Density:1,100
  • Storage Temperature:Raumtemperatur
Specifications

Specifications

  • Packung:
    Glass, amber
  • VE:
    5 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.