Express Eingabe
Austria
AuftragsverfolgungKontakt
Reagent Reservoirs

Reagent Reservoirs

96 wells, 195 ml, 127,8×85,5×31,4 mm, -, 5 packs of 10 each
Artikelnr. SIMPR225-7
Anbieter:   Simport Scientific
Reagent Reservoirs
Artikelnr. SIMPR225-7
Anbieter:  Simport Scientific
Reagent Reservoirs
Reagent Reservoirs

96 wells, 195 ml, 127,8×85,5×31,4 mm, -, 5 packs of 10 each

Preis
690,00 €
Einloggen für Verfügbarkeit
1 * 50 ST
Product Details & Documents

For labs involved with fraction collecting, compound storage, plate replication, and mixing applications. Suitable for a range of liquid handling configurations from 8 or 12 channel pipettes to 96 or 384 pipette heads.

  • Robot friendly, designed for simple, economical handling of reagents in automated systems
  • Bottom designed to enable funneling of liquid, maximising reagent recovery with pipettes
  • Low-profile
  • Chemical and heat resistant, can be stored at temperatures as low as –80 °C
  • Autoclavable
  • Can be gamma irradiated
  • Certified DNase/RNase and pyrogen free

Ideal for small volume robotic tips used in high throughput instruments. Choose from a single reservoir or partitioned models for several liquids.

Conforms to ANSI/SLAS standard dimensions, they are compatible with virtually any automated liquid handler, stacker, and manual procedure.

Made from natural virgin polypropylene, providing a low binding surface to prevent samples from sticking to the sidewalls during elution and is chemically inert for combinatorial chemistry applications.

Spezifikationen
  • Verpackt:
    5 packs of 10 each
  • Steril:
    -
  • Typ:
    96 wells
  • Kapazität:
    195 ml
  • Seq:
    04
  • B×T×H:
    127,8×85,5×31,4 mm