Reagent Reservoirs
127,8×85×9 mm, -, 5 packs of 10 each
SIMPR225-15
For labs involved with fraction collecting, compound storage, plate replication, and mixing applications. Suitable for a range of liquid handling configurations from 8 or 12 channel pipettes to 96 or 384 pipette heads.
- Robot friendly, designed for simple, economical handling of reagents in automated systems
- Bottom designed to enable funneling of liquid, maximising reagent recovery with pipettes
- Low-profile
- Chemical and heat resistant, can be stored at temperatures as low as –80 °C
- Autoclavable
- Can be gamma irradiated
- Certified DNase/RNase and pyrogen free
Ideal for small volume robotic tips used in high throughput instruments. Choose from a single reservoir or partitioned models for several liquids.
Conforms to ANSI/SLAS standard dimensions, they are compatible with virtually any automated liquid handler, stacker, and manual procedure.
Made from natural virgin polypropylene, providing a low binding surface to prevent samples from sticking to the sidewalls during elution and is chemically inert for combinatorial chemistry applications.
- Verpackt:5 packs of 10 each
- Steril:-
- Seq:06
- B×T×H:127,8×85×9 mm



