Express Eingabe
Austria
AuftragsverfolgungKontakt
Reagent Reservoirs | Avantor

Reagent Reservoirs

1 well, 190 ml, 127,8×85,5×31,4 mm, -, 5 packs of 10 each
Artikelnr. SIMPR225-1
Anbieter:   Simport Scientific
Reagent Reservoirs
Artikelnr. SIMPR225-1
Anbieter:  Simport Scientific
Reagent Reservoirs
Reagent Reservoirs

1 well, 190 ml, 127,8×85,5×31,4 mm, -, 5 packs of 10 each

Preis
690,00 €
Einloggen für Verfügbarkeit
1 * 50 ST
Product Details & Documents

For labs involved with fraction collecting, compound storage, plate replication, and mixing applications. Suitable for a range of liquid handling configurations from 8 or 12 channel pipettes to 96 or 384 pipette heads.

  • Robot friendly, designed for simple, economical handling of reagents in automated systems
  • Bottom designed to enable funneling of liquid, maximising reagent recovery with pipettes
  • Low-profile
  • Chemical and heat resistant, can be stored at temperatures as low as –80 °C
  • Autoclavable
  • Can be gamma irradiated
  • Certified DNase/RNase and pyrogen free

Ideal for small volume robotic tips used in high throughput instruments. Choose from a single reservoir or partitioned models for several liquids.

Conforms to ANSI/SLAS standard dimensions, they are compatible with virtually any automated liquid handler, stacker, and manual procedure.

Made from natural virgin polypropylene, providing a low binding surface to prevent samples from sticking to the sidewalls during elution and is chemically inert for combinatorial chemistry applications.

Spezifikationen
  • Verpackt:
    5 packs of 10 each
  • Steril:
    -
  • Typ:
    1 well
  • Kapazität:
    190 ml
  • Seq:
    01
  • B×T×H:
    127,8×85,5×31,4 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.