ContactUsLinkComponent
crystal clear, 300 strips per case Made of a special polymer with improved optical properties leading to high transmission rates ideally suited for small samples with low signal intensity Reduced shrinking during heating and cooling phases very tig 1 * 300 ST
Artikelnr. FTIT4TI-0755
Lieferant:  4titude Ltd.
crystal clear, 300 strips per case Made of a special polymer with improved optical properties leading to high transmission rates ideally suited for small samples with low signal intensity Reduced shrinking during heating and cooling phases very tig 1 * 300 ST
Artikelnr. FTIT4TI-0755
Lieferant:  4titude Ltd.
Frequently Bought Together