ContactUsLinkComponent
External Dimensions: 460 x 460 x 460 mm Internal Dimensions: 320 x 320 x 320 mm Dry-Ice Mass: 19.1 kg Payload Volume: 11.8 L Duration: 96+ 1 * 1 ST
Artikelnr. DGINFRAPHT30
Lieferant:  DGP INTELSIUS
External Dimensions: 460 x 460 x 460 mm Internal Dimensions: 320 x 320 x 320 mm Dry-Ice Mass: 19.1 kg Payload Volume: 11.8 L Duration: 96+ 1 * 1 ST
Artikelnr. DGINFRAPHT30
Lieferant:  DGP INTELSIUS
Frequently Bought Together