Containers with Sloping Base
Hygienic valve kit with silicone seals, —
Artikelnr. SAMPA136-01S
Anbieter: Sampling Systems
Springe zu:
Product Details & Documents
These drums which incorporate a sloping base allow the container to drain completely.
- Made from 316L stainless steel
- Includes lid, clamp and silicone seal
- Other seals available
- Ferrule for easy emptying
- Handles enable easy transportation
The Sampling Systems range of containers with sloping base ensures that material can be completely drained from within and include a ferrule at the base for easy emptying. They have a mirror polish finish for easy cleaning.
Spezifikationen
- Kapazität:—
- Bezeichnung:Hygienic valve kit with silicone seals
- Seq:06