Express Eingabe
Austria
AuftragsverfolgungKontakt
Human IL-4 ELISpot Kit

Human IL-4 ELISpot Kit

Human IL-4 ELISpot kit, 5×96 tests
Artikelnr. ABCAAB281966-5X96
Anbieter:   Abcam
Human IL-4 ELISpot Kit
Artikelnr. ABCAAB281966-5X96
Anbieter:  Abcam
CAS Nummer:  
Human IL-4 ELISpot Kit
Human IL-4 ELISpot Kit

Human IL-4 ELISpot kit, 5×96 tests

CAS Nummer:  
Preis
1 210,00 €
Einloggen für Verfügbarkeit
1 * 480 Tests
Product Details & Documents

Human IL-4 ELISpot Kit is a ELISpot Kit for the sensitive detection of Human IL-4 production in a single cell suspension.

Spezifikationen

Spezifikationen

  • Bezeichnung:
    Human IL-4 ELISpot kit
  • Tests pro Kit:
    5×96 tests
  • Cat. No.:
    ABCAAB281966-5X96
  • Lagertemperatur:
    4 °C
  • Species reactivity:
    Human
  • Probentyp:
    Suspension cells
  • Processing time:
    Multiple steps
  • Antigen-Symbol:
    IL-4
  • Regulatory status:
    RUO

Spezifikationen

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.