Human IL-4 ELISpot Kit
Human IL-4 ELISpot kit, 96 tests
Artikelnr. ABCAAB281966-1X96
Anbieter: Abcam
Springe zu:
Product Details & Documents
Human IL-4 ELISpot Kit is a ELISpot Kit for the sensitive detection of Human IL-4 production in a single cell suspension.
Spezifikationen
Spezifikationen
- Bezeichnung:Human IL-4 ELISpot kit
- Tests pro Kit:96 tests
- Cat. No.:ABCAAB281966-1X96
- Lagertemperatur:4 °C
- Species reactivity:Human
- Probentyp:Suspension cells
- Processing time:Multiple steps
- Antigen-Symbol:IL-4
- Regulatory status:RUO



