Masterflex® Fitting, Nylon, Straight, Male Luer to Hose Barb Adapter, 1/8" ID; 25/PK (use with Separate Lock Ring)
Artikelnr. MFLX45505-44
Lieferant: Avantor Fluid Handling
CE certificate
Avantor exclusive
Über diesen Artikel
Stellen Sie Ihre Prozessintegrität mit vollständiger Materialrückverfolgbarkeit sicher.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Hose barbs are available from ¹/₁₆" to ¹/₄" with either male or female luer connections. Male luer connections are available with either integral lock ring or separate lock ring.
- Tüllengröße/Schlauch Innen-Ø (Port 1):⅛"
- Verbindungstyp:Male luer to barb
- Flow pattern:Straight
- Bezeichnung:Luer mit Außenkegel x 1/8“-Schlauchnippel (für separaten Verschlussring), Nylon, 25/Pck
- Seq:16
- Environmentally Preferable:
- Modell:45505-44
- Connection type (port 1):Male luer slip
- No. of ports:2
- Garantie:3 Months
- VE:25
- Fitting material:Nylon
Frequently Bought Together