AuftragsverfolgungKontakt
CytoFLEX SRT Software Upgrades | Avantor
CytoFLEX SRT Software Upgrades
Activation key to upgrade the CytoFLEX SRT V0-B2-Y5-R0 Cell Sorter with additional parameters. Upon installation the instrument will have 3 active lasers (405 nm, 488 nm, 561 nm) and 12 fluorescence detectors in the V5-B2-Y5-R0 configuration
Artikelnr. BECMC65144
Anbieter:   Beckman Coulter
CytoFLEX SRT Software Upgrades
Artikelnr. BECMC65144
Anbieter:  Beckman Coulter
CytoFLEX SRT Software Upgrades
Activation key to upgrade the CytoFLEX SRT V0-B2-Y5-R0 Cell Sorter with additional parameters. Upon installation the instrument will have 3 active lasers (405 nm, 488 nm, 561 nm) and 12 fluorescence detectors in the V5-B2-Y5-R0 configuration
Preis
Preis anfragen
Einloggen für Verfügbarkeit
1 * 1 ST
Product Details & Documents
Spezifikationen
  • Bezeichnung:
    Activation key to upgrade the CytoFLEX SRT V0-B2-Y5-R0 Cell Sorter with additional parameters. Upon installation the instrument will have 3 active lasers (405 nm, 488 nm, 561 nm) and 12 fluorescence detectors in the V5-B2-Y5-R0 configuration
  • Seq:
    8