ContactUsLinkComponent
Degassing Channel Tubing: Systec AF™ (0.045’’ ID) Maximum Channel Pressure: 70 PSI) Wetted Materials: Systec AF™, PEEK, Glass-filled PTFE Liquid Connection: 1/4’’-28 UNF threaded flat-bottom port Size (L x H x W): 245 x 111 x 59 mm 1 * 1 ST
Artikelnr. HPLC0003-6622
Degassing Channel Tubing: Systec AF™ (0.045’’ ID) Maximum Channel Pressure: 70 PSI) Wetted Materials: Systec AF™, PEEK, Glass-filled PTFE Liquid Connection: 1/4’’-28 UNF threaded flat-bottom port Size (L x H x W): 245 x 111 x 59 mm 1 * 1 ST
Artikelnr. HPLC0003-6622
Frequently Bought Together