Masterflex® Spray Gun, PTFE for DI Water, 1/2" NPT(F); 1/Ea
Artikelnr. MFLX98515-03
Lieferant: Avantor Fluid Handling
Über diesen Artikel
No splashing.
- Recirculating gun has a contamination free fluid path to eliminate bacterial growth
- Highly resistant to corrosive substances and fumes
- Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi
These Masterflex® Tank Dispensing Spray Guns feature a special tip to eliminate splashing. Choose from PTFE or stainless steel construction for a wide range of applications.
- Bezeichnung:Spray gun, PTFE for DI water; 1/2"
- Environmentally Preferable:
- Wetted materials:PTFE
- Typ:98515-03
- Garantie:1 year
- Connection type:NPT(F)
- VE:1
Frequently Bought Together