In-line Filter 70um PVDF for Dosing Pumps 78167-08 to -10
Catalog # MFLX78167-52
Supplier: Avantor Fluid Handling
1 year warranty
About this item
Replacement parts keep your dosing pump running smoothly.
- Opimize performance of your KNF Remote-Control Diaphragm Liquid Dosing Pumps
For use with KNF Remote-Control Diaphragm Liquid Dosing Pumps, pumps sold separately.
- Bezeichnung:In-line Filter 70um PVDF for Dosing Pumps 78167-08 to -10
- Environmentally Preferable:
- Wetted materials:PVDF
- Garantie:1 year
- VE:1
Frequently Bought Together