Masterflex® Fitting, Polypropylene, Straight, Female Luer to Thread Adapter, with 5/16" Hex to 1/4-28 UNF; 25/PK
Artikelnr. MFLX45508-66
Lieferant: Avantor Fluid Handling
CE certificate
Über diesen Artikel
Leichter Übergang zwischen Luer-Fittings und UNF- oder NPT-Gewinde.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Diese Adapter biete einen Übergang zwischen Luer-Fittings und UNF- oder NPT-Gewinde. Außen-Luer- wie Innen-Luer-Verbindungen sind verfügbar. Alle Gewindeverbindungen sind Innengewinde.
- Verbindungstyp:Female Luer to Thread
- Flow pattern:Straight
- Bezeichnung:Innen-Luer x 1/4-28 UNF-Gewinde, PP, 25/Pck 45508-66
- Seq:05
- Environmentally Preferable:
- Modell:45508-66
- Connection type (port 1):Female luer
- Connection type (port 2):1/4-28 UNF thread
- No. of ports:2
- VE:25
- Fitting material:Polypropylene
- Thread size UNF(M) (port 1):1/4"-28
Frequently Bought Together