Masterflex® Fitting, Polycarbonate, Straight, Male Luer x Female Luer; 25/PK
Catalog # MFLX45501-82
Supplier: Avantor Fluid Handling
CE certificate
Avantor exclusive
Specifications
See all product configurations- Catalog No:
- MFLX45501-82
- MFLX45501-28
- MFLX45501-22
- MFLX45501-64
- MFLX45504-56
- MFLX45501-84
- MFLX45501-56
- MFLX30526-10
- Modell:
- 45501-82
- 45501-28
- 45501-22
- 45501-64
- 45504-56
- 45501-84
- 45501-56
- 30526-10
- Connection type (port 1):
- Female luer
- Female luer
- Female luer
- Female luer
- Male luer lock
- Female luer
- Female luer
- Female luer
- Connection type (port 2):
- Male luer slip
- Cap
- Female luer
- Cap with tether loop
- Plug
- Male luer slip
- Female luer
- Male luer lock
- Connection type (port 3):
- Female luer
- No. of ports:
- 2
- 1
- 2
- 1
- 1
- 2
- 3
- 2
- Garantie:
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- 3 Months
- Out of box
- Currently Viewing
Specifications
About this item
Einfacher Übergang auf ein Luer-Fitting.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Vervollständigen Sie Ihre Luer-Anschlusssysteme ganz einfach mit diesen Premium-Anschlüssen. Im Lieferumfang sind einige Sonderkonfigurationen enthalten, die nur in Polycarbonat erhältlich sind. Alle weiblichen Anschlüsse können entweder mit Slip- oder Luer-Lock-Anschlüssen verwendet werden. Stopfen und Kappen sind ebenfalls erhältlich. Material aus Polycarbonat weist eine gute chemische Kompatibilität auf und kann durch Ethylenoxid- oder Gammabestrahlung sterilisiert werden.
- Verbindungstyp:Female Luer to Male Luer
- Flow pattern:Straight
- Bezeichnung:Außen-Luer x Innen-Luer, PC, 25/Pk
- Seq:12
- Environmentally Preferable:
- Modell:45501-82
- Connection type (port 1):Female luer
- Connection type (port 2):Male luer slip
- No. of ports:2
- Garantie:3 Months
- VE:25
- Fitting material:Polycarbonate
Frequently Bought Together